2-Bromo-1-iodo-3-(trifluoromethoxy)benzene 95%

Lieferant: Apollo Scientific

PC50968-1G PC50968-250MG PC50968-5G
APOSPC50968-1GEA 429 EUR
APOSPC50968-1G APOSPC50968-250MG APOSPC50968-5G
2-Bromo-1-iodo-3-(trifluoromethoxy)benzene 95%
2-Bromo-1-iodo-3-(trifluoromethoxy)benzene
Formel: C₇H₃BrF₃IO
Molecular Weight: 366,902 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18398074
CAS-Nummer: 1261752-43-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR