Pro-Gly-OMe TFA

Lieferant: Apollo Scientific

PC510015-1G PC510015-5G
APOSPC510015-1GEA 306 EUR
APOSPC510015-1G APOSPC510015-5G
Pro-Gly-OMe TFA
Pro-Gly-OMe TFA
Formel: C₈H₁₄N₂O₃·CF₃CO₂H
Molecular Weight: 300,232 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16495871
CAS-Nummer: 634922-10-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR