3-(4-Fluorophenyl)-3-oxo-2-(4-pyridinyl)propanenitrile
Lieferant: Apollo Scientific
PC510022-1G
PC510022-5G
APOSPC510022-1GEA
397
EUR
APOSPC510022-1G
APOSPC510022-5G
3-(4-Fluorophenyl)-3-oxo-2-(4-pyridinyl)propanenitrile
3-(4-Fluorophenyl)-3-oxo-2-(4-pyridinyl)propanenitrile
Formel:
C₁₄H₉FN₂O Molecular Weight: 240,232 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18071009 CAS-Nummer: 148671-42-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...