1-Boc-4-fluoro-1H-indazole

Lieferant: Apollo Scientific

PC510038-1G PC510038-5G
APOSPC510038-1GEA 337 EUR
APOSPC510038-1G APOSPC510038-5G
1-Boc-4-fluoro-1H-indazole
1-Boc-4-fluoro-1H-indazole
Formel: C₁₂H₁₃FN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18911747
CAS-Nummer: 1305320-65-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR