(R)-N-Boc-3-(4-fluorophenoxy)pyrrolidine
Lieferant: Apollo Scientific
PC510049-1G
PC510049-5G
APOSPC510049-1GEA
397
EUR
APOSPC510049-1G
APOSPC510049-5G
(R)-N-Boc-3-(4-fluorophenoxy)pyrrolidine
(R)-N-Boc-3-(4-fluorophenoxy)pyrrolidine
Formel:
C₁₅H₂₀FNO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD19442783 CAS-Nummer: 1314419-66-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...