3-Fluoro-4-(methylsulphonamido)benzoic acid

Lieferant: Apollo Scientific

PC510056-1G PC510056-5G
APOSPC510056-1GEA 275 EUR
APOSPC510056-1G APOSPC510056-5G
3-Fluoro-4-(methylsulphonamido)benzoic acid
3-Fluoro-4-(methylsulphonamido)benzoic acid
Formel: C₈H₈FNO₄S
Molecular Weight: 233.217 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18285558
CAS-Nummer: 716361-59-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR