1-(Boc-amino)-3-bromo-1-(4-fluorophenyl)propane
Lieferant: Apollo Scientific
PC510079-1G
PC510079-5G
APOSPC510079-1GEA
499
EUR
APOSPC510079-1G
APOSPC510079-5G
1-(Boc-amino)-3-bromo-1-(4-fluorophenyl)propane
1-(Boc-amino)-3-bromo-1-(4-fluorophenyl)propane
Formel:
C₁₄H₁₉BrFNO₂ Molecular Weight: 332.209 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06410615 CAS-Nummer: 924817-75-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...