5-[2-(Trifluoromethoxy)phenyl]-3-pyridinecarbaldehyde

Lieferant: Apollo Scientific

PC510081-1G PC510081-5G
APOSPC510081-1GEA 397 EUR
APOSPC510081-1G APOSPC510081-5G
5-[2-(Trifluoromethoxy)phenyl]-3-pyridinecarbaldehyde
5-[2-(Trifluoromethoxy)phenyl]-3-pyridinecarbaldehyde
Formel: C₁₃H₈F₃NO₂
Molecular Weight: 267,203 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410236
CAS-Nummer: 887973-92-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR