5-Fluoro-1-methyl-1H-indazole-6-boronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Borono-5-fluoro-1-methyl-1H-indazole, (5-Fluoro-1-methyl-1H-indazol-6-yl)boronic acid

PC51013-1G PC51013-250MG PC51013-100MG
APOSPC51013-1GEA 931 EUR
APOSPC51013-1G APOSPC51013-250MG APOSPC51013-100MG
5-Fluoro-1-methyl-1H-indazole-6-boronic acid
5-Fluoro-1-methyl-1H-indazole-6-boronic acid
Formel: C₈H₈BFN₂O₂
Molecular Weight: 193.971 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18089818
CAS-Nummer: 2096331-06-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR