5-Bromo-1,1,1,2,2-pentafluoropentane

Lieferant: Apollo Scientific
Achtung

PC510167-1G PC510167-250MG
APOSPC510167-1GEA 754 EUR
APOSPC510167-1G APOSPC510167-250MG
5-Bromo-1,1,1,2,2-pentafluoropentane
5-Bromo-1,1,1,2,2-pentafluoropentane
Formel: C₅H₆BrF₅
Molecular Weight: 240,997 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27935381
CAS-Nummer: 1383437-60-2
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR