5-Fluoro-1-methyl-1H-indazole-6-carboxaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Fluoro-1-methyl-1H-indazole-6-carbaldehyde, 5-Fluoro-6-formyl-1-methyl-1H-indazole

PC51018-100MG PC51018-1G PC51018-500MG
APOSPC51018-100MGEA 217 EUR
APOSPC51018-100MG APOSPC51018-1G APOSPC51018-500MG
5-Fluoro-1-methyl-1H-indazole-6-carboxaldehyde
5-Fluoro-1-methyl-1H-indazole-6-carboxaldehyde
Formel: C₉H₇FN₂O
Molecular Weight: 178,163 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18384870
CAS-Nummer: 1286734-88-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR