N-Boc-3-amino-2,2-difluoropropionic acid
Lieferant: Apollo Scientific
PC510199-1G
PC510199-250MG
APOSPC510199-1GEA
713
EUR
APOSPC510199-1G
APOSPC510199-250MG
N-Boc-3-amino-2,2-difluoropropionic acid
N-Boc-3-amino-2,2-difluoropropionic acid
Formel:
C₈H₁₃F₂NO₄ Molecular Weight: 225.19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13189573 CAS-Nummer: 1196145-07-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...