3-Bromo-5-(trifluoromethyl)pyrazole

Lieferant: Apollo Scientific
Achtung

PC510204-1G PC510204-250MG
APOSPC510204-1GEA 0 EUR
APOSPC510204-1G APOSPC510204-250MG
3-Bromo-5-(trifluoromethyl)pyrazole
3-Bromo-5-(trifluoromethyl)pyrazole
Formel: C₄H₂BrF₃N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD12755885
CAS-Nummer: 93608-11-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR