1-(2-Chloroethyl)-2-fluorobenzene
Lieferant: Apollo Scientific
PC510242-1G
PC510242-250MG
PC510242-5G
APOSPC510242-1GEA
478
EUR
APOSPC510242-1G
APOSPC510242-250MG
APOSPC510242-5G
1-(2-Chloroethyl)-2-fluorobenzene
1-(2-Chloroethyl)-2-fluorobenzene
Formel:
C₈H₈ClF Molecular Weight: 158,601 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11110026 CAS-Nummer: 119779-12-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...