2-Chloro-7-(trifluoromethoxy)quinoline-3-carbaldehyde

Lieferant: Apollo Scientific

PC510262-1G PC510262-5G PC510262-250MG
APOSPC510262-1GEA 601 EUR
APOSPC510262-1G APOSPC510262-5G APOSPC510262-250MG
2-Chloro-7-(trifluoromethoxy)quinoline-3-carbaldehyde
2-Chloro-7-(trifluoromethoxy)quinoline-3-carbaldehyde
Formel: C₁₁H₅ClF₃NO₂
Molecular Weight: 275,611 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23132257
CAS-Nummer: 1254366-15-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR