5-Bromo-2-(difluoromethoxy)pyridine 95%

Lieferant: Apollo Scientific
Achtung

PC510286-1G PC510286-250MG PC510286-5G
APOSPC510286-1GEA 148 EUR
APOSPC510286-1G APOSPC510286-250MG APOSPC510286-5G
5-Bromo-2-(difluoromethoxy)pyridine 95%
5-Bromo-2-(difluoromethoxy)pyridine
Formel: C₆H₄BrF₂NO
Molecular Weight: 224.003 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11977440
CAS-Nummer: 899452-26-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR