Methyl 5-bromo-6-fluoroindole-2-carboxylate

Lieferant: Apollo Scientific

PC510303-1G PC510303-250MG PC510303-5G
APOSPC510303-1GEA 601 EUR
APOSPC510303-1G APOSPC510303-250MG APOSPC510303-5G
Methyl 5-bromo-6-fluoroindole-2-carboxylate
Methyl 5-bromo-6-fluoroindole-2-carboxylate
Formel: C₁₀H₇BrFNO₂
Molecular Weight: 272,07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29055290
CAS-Nummer: 2006277-60-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR