2,2-Difluoro-2-[4-(trifluoromethyl)phenyl]acetic acid

Lieferant: Apollo Scientific
Gefahr

PC510326-5G PC510326-1G
APOSPC510326-5GEA 1010 EUR
APOSPC510326-5G APOSPC510326-1G
2,2-Difluoro-2-[4-(trifluoromethyl)phenyl]acetic acid
2,2-Difluoro-2-[4-(trifluoromethyl)phenyl]acetic acid
Formel: C₉H₅F₅O₂
Molecular Weight: 240,127 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16756774
CAS-Nummer: 73790-11-1
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR