α,α-Difluoro-2-(4-methoxyphenyl)acetic acid
Lieferant: Apollo Scientific
Synonyme:
2,2-Difluoro-2-(4-methoxyphenyl)acetic acid
PC510343-1G
PC510343-250MG
PC510343-5G
APOSPC510343-1GEA
374
EUR
APOSPC510343-1G
APOSPC510343-250MG
APOSPC510343-5G
α,α-Difluoro-2-(4-methoxyphenyl)acetic acid
α,α-Difluoro-2-(4-methoxyphenyl)acetic acid
Formel:
C₉H₈F₂O₃ Molecular Weight: 202.155 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11007700 CAS-Nummer: 1027513-97-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...