2,6-Dichloro-5-fluoropyrimidine-4-carboxamide

Lieferant: Apollo Scientific

PC510345-1G PC510345-250MG
APOSPC510345-1GEA 611 EUR
APOSPC510345-1G APOSPC510345-250MG
2,6-Dichloro-5-fluoropyrimidine-4-carboxamide
2,6-Dichloro-5-fluoropyrimidine-4-carboxamide
Formel: C₅H₂Cl₂FN₃O
Molecular Weight: 209,993 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29065912
CAS-Nummer: 2006277-92-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR