4-Bromo-2,6-difluoro-α-methylbenzyl alcohol 95%

Lieferant: Apollo Scientific

Synonyme: 4-Bromo-2,6-difluoro-.alpha.-methyl-benzenemethanol, 1-(4-Bromo-2,6-difluorophenyl)ethanol

PC510353-5G
APOSPC510353-5GEA 101 EUR
APOSPC510353-5G
4-Bromo-2,6-difluoro-α-methylbenzyl alcohol 95%
4-Bromo-2,6-difluoro-α-methylbenzyl alcohol
Formel: C₈H₇BrF₂O
Molecular Weight: 237,041 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25459281
CAS-Nummer: 1214900-62-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR