4-Chloro-6-fluoro-2-(trichloromethyl)quinoline

Lieferant: Apollo Scientific

PC51037-1G PC51037-5G
APOSPC51037-1GEA 0 EUR
APOSPC51037-1G APOSPC51037-5G
4-Chloro-6-fluoro-2-(trichloromethyl)quinoline
4-Chloro-6-fluoro-2-(trichloromethyl)quinoline
Formel: C₁₀H₄Cl₄FN
Lagertemperatur: Raumtemperatur
MDL: MFCD02684139
CAS-Nummer: 927800-47-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR