Perfluoro(1,2-dibromo-3,4-dichlorobutane)
Lieferant: Apollo Scientific
PC51061-1G
PC51061-5G
APOSPC51061-1GEA
136
EUR
APOSPC51061-1G
APOSPC51061-5G
Perfluoro(1,2-dibromo-3,4-dichlorobutane)
Perfluoro(1,2-dibromo-3,4-dichlorobutane)
Formel:
C₄Br₂Cl₂F₆ Siedepunkt: 63 °C (14 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD19442561 CAS-Nummer: 375-27-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...