2-Bromo-2-fluoropropanoic acid
Lieferant: Apollo Scientific
Synonyme:
2-Bromo-2-fluoropropionic acid
PC51071-1G
PC51071-250MG
APOSPC51071-1GEA
535
EUR
APOSPC51071-1G
APOSPC51071-250MG
2-Bromo-2-fluoropropanoic acid
2-Bromo-2-fluoropropanoic acid
Formel:
C₃H₄BrFO₂ Molecular Weight: 170,965 g/mol Siedepunkt: 99 °C (1 mm Hg) Dichte: 1,811 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD19442053 CAS-Nummer: 132629-22-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...