3-Fluorophenylsulphur pentafluoride

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Fluoro-3-(pentafluorothio)benzene, 1-Fluoro-3-(pentafluorosulphanyl)benzene

PC51096-1G PC51096-5G
APOSPC51096-1GEA 145 EUR
APOSPC51096-1G APOSPC51096-5G
3-Fluorophenylsulphur pentafluoride
3-Fluorophenylsulphur pentafluoride
Formel: C₆H₄F₆S
Molecular Weight: 222,151 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21362225
CAS-Nummer: 1422-41-9
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR