2-Aminophenylsulphur pentafluoride

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Pentafluorosulphanyl)aniline, 2-(Pentafluorothio)aniline

PC51097-1G PC51097-5G
APOSPC51097-1GEA 422 EUR
APOSPC51097-1G APOSPC51097-5G
2-Aminophenylsulphur pentafluoride
2-Aminophenylsulphur pentafluoride
Formel: C₆H₆F₅NS
Molecular Weight: 219,176 g/mol
Schmelzpunkt: 28,8…30.5 °C
Lagertemperatur: Kühlschrank
MDL: MFCD21362226
CAS-Nummer: 1246998-10-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR