Fluoromalonaldehyde, sodium salt
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Sodium 2-fluoro-3-oxoprop-1-en-1-olate, Fluoropropane-1,3-dial sodium salt
PC51104-1G
PC51104-250MG
APOSPC51104-1GEA
319
EUR
APOSPC51104-1G
APOSPC51104-250MG
Fluoromalonaldehyde, sodium salt
Fluoromalonaldehyde, sodium salt
Formel:
C₃H₂FO₂·Na Molecular Weight: 112,035 g/mol Schmelzpunkt: >200 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD21362228 CAS-Nummer: 29548-72-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...