2-Chloro-3-fluoro-4-hydroxypyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-3-fluoropyridin-4-ol

PC51119-100MG PC51119-1G PC51119-5G
APOSPC51119-100MGEA 65.4 EUR
APOSPC51119-100MG APOSPC51119-1G APOSPC51119-5G
2-Chloro-3-fluoro-4-hydroxypyridine
2-Chloro-3-fluoro-4-hydroxypyridine
Formel: C₅H₃ClFNO
Molecular Weight: 147.535 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13189885
CAS-Nummer: 1184172-46-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR