2-Amino-5-bromo-4-fluorobenzoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-2-carboxy-5-fluoroaniline, 5-Bromo-4-fluoroanthranilic acid

PC51135-25G PC51135-100G
APOSPC51135-25GEA 102 EUR
APOSPC51135-25G APOSPC51135-100G
2-Amino-5-bromo-4-fluorobenzoic acid
2-Amino-5-bromo-4-fluorobenzoic acid
Formel: C₇H₅BrFNO₂
Molecular Weight: 234.023 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06409112
CAS-Nummer: 143945-65-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR