2-Fluoro-6-methylbenzyl alcohol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (6-Fluoro-o-tolyl)methanol, (2-Fluoro-6-methylphenyl)methanol

PC51146-1G
APOSPC51146-1GEA 56.7 EUR
APOSPC51146-1G
2-Fluoro-6-methylbenzyl alcohol 97%
2-Fluoro-6-methylbenzyl alcohol
Formel: C₈H₉FO
Molecular Weight: 140.155 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06660280
CAS-Nummer: 478163-35-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR