2-Amino-3,4-difluorobenzonitrile 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Cyano-2,3-difluoroaniline, 3,4-Difluoroanthranilonitrile

PC51147-5G PC51147-1G
APOSPC51147-5GEA 81.2 EUR
APOSPC51147-5G APOSPC51147-1G
2-Amino-3,4-difluorobenzonitrile 99%
2-Amino-3,4-difluorobenzonitrile
Formel: C₇H₄F₂N₂
Molecular Weight: 154,117 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD21606584
CAS-Nummer: 1384265-40-0
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR