3-(Pentafluorothio)phenylacetylene 96%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-Ethynyl-3-(pentafluorosulphanyl)benzene, 3-Ethynylphenylsulphur pentafluoride

PC51148-1G PC51148-250MG PC51148-5G
APOSPC51148-1GEA 422 EUR
APOSPC51148-1G APOSPC51148-250MG APOSPC51148-5G
3-(Pentafluorothio)phenylacetylene 96%
3-(Pentafluorothio)phenylacetylene
Formel: C₈H₅F₅S
Lagertemperatur: Kühlschrank
MDL: MFCD23166761
CAS-Nummer: 1426290-10-9
UN: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR