3-Bromo-4-hydroxyphenylsulphur pentafluoride
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Bromo-4-(pentafluorothio)phenol, 2-Bromo-4-(pentafluorosulphanyl)phenol
PC51151-1G
PC51151-250MG
APOSPC51151-1GEA
464
EUR
APOSPC51151-1G
APOSPC51151-250MG
3-Bromo-4-hydroxyphenylsulphur pentafluoride
3-Bromo-4-hydroxyphenylsulphur pentafluoride
Formel:
C₆H₄BrF₅OS Molecular Weight: 299.056 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23166763 CAS-Nummer: 1426290-12-1 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...