4-Bromo-5-fluoro-2-nitrophenol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-4-fluoro-2-hydroxynitrobenzene

PC51153-1G PC51153-5G PC51153-25G
APOSPC51153-1GEA 24.8 EUR
APOSPC51153-1G APOSPC51153-5G APOSPC51153-25G
4-Bromo-5-fluoro-2-nitrophenol 97%
4-Bromo-5-fluoro-2-nitrophenol
Formel: C₆H₃BrFNO₃
Molecular Weight: 235.995 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14583144
CAS-Nummer: 1016234-87-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR