5'-Bromo-2,2,2,2'-tetrafluoroacetophenone 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(5-Bromo-2-fluorophenyl)-2,2,2-trifluoroethanone, 1-(5-Bromo-2-fluorophenyl)-2,2,2-trifluoroethan-1-one
PC51159-1G
PC51159-250MG
PC51159-5G
APOSPC51159-1GEA
255
EUR
APOSPC51159-1G
APOSPC51159-250MG
APOSPC51159-5G
5'-Bromo-2,2,2,2'-tetrafluoroacetophenone 95%
5'-Bromo-2,2,2,2'-tetrafluoroacetophenone
Formel:
C₈H₃BrF₄O Molecular Weight: 271.006 g/mol Siedepunkt: 53 °C (1 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD12547807 CAS-Nummer: 617706-15-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...