2-Bromo-4-(difluoromethyl)thiophene
Lieferant: Apollo Scientific
|
Achtung
|
PC51160-1G
PC51160-5G
PC51160-250MG
APOSPC51160-1GEA
219
EUR
APOSPC51160-1G
APOSPC51160-5G
APOSPC51160-250MG
2-Bromo-4-(difluoromethyl)thiophene
2-Bromo-4-(difluoromethyl)thiophene
Formel:
C₅H₃BrF₂S Molecular Weight: 213.043 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD23166764 CAS-Nummer: 1426290-09-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...