4-Ethynyl-2-fluorobenzaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Fluoro-4-formylphenylacetylene

PC51170-1G PC51170-250MG PC51170-5G
APOSPC51170-1GEA 427 EUR
APOSPC51170-1G APOSPC51170-250MG APOSPC51170-5G
4-Ethynyl-2-fluorobenzaldehyde
4-Ethynyl-2-fluorobenzaldehyde
Formel: C₉H₅FO
Molecular Weight: 148.134 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD23703632
CAS-Nummer: 1267502-64-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR