2-Nitro-4-[(trifluoromethyl)sulphonyl]phenol

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Hydroxy-3-nitrophenyl trifluoromethyl sulphone, 2-Nitro-4-((trifluoromethyl)sulphonyl)phenol, 2-Hydroxy-5-[(trifluoromethyl)sulphonyl]nitrobenzene

PC51177-1G PC51177-250MG PC51177-5G
APOSPC51177-1GEA 179 EUR
APOSPC51177-1G APOSPC51177-250MG APOSPC51177-5G
2-Nitro-4-[(trifluoromethyl)sulphonyl]phenol
2-Nitro-4-[(trifluoromethyl)sulphonyl]phenol
Formel: C₇H₄F₃NO₅S
Molecular Weight: 271,171 g/mol
Schmelzpunkt: 58…61 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00986405
CAS-Nummer: 15183-75-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR