5-Bromo-3-fluoro-2-hydroxybenzoic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-2-carboxy-6-fluorophenol, 5-Bromo-3-fluorosalicylic acid

PC51186-1G PC51186-250MG PC51186-5G
APOSPC51186-1GEA 130 EUR
APOSPC51186-1G APOSPC51186-250MG APOSPC51186-5G
5-Bromo-3-fluoro-2-hydroxybenzoic acid 97%
5-Bromo-3-fluoro-2-hydroxybenzoic acid
Formel: C₇H₄BrFO₃
Molecular Weight: 235,007 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06203615
CAS-Nummer: 251300-29-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR