4-[(4-Methylpiperazin-1-yl)methyl]-3-(trifluoromethyl)aniline 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-((4-Methylpiperazin-1-yl)methyl)-3-(trifluoromethyl)aniline

PC51192-5G PC51192-25G
APOSPC51192-5GEA 69.6 EUR
APOSPC51192-5G APOSPC51192-25G
4-[(4-Methylpiperazin-1-yl)methyl]-3-(trifluoromethyl)aniline 95+%
4-[(4-Methylpiperazin-1-yl)methyl]-3-(trifluoromethyl)aniline
Formel: C₁₃H₁₈F₃N₃
Molecular Weight: 273,297 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09909924
CAS-Nummer: 694499-26-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR