4-Fluoro-2-methyl-5-nitrophenol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Fluoro-2-hydroxy-4-nitrotoluene, 4-Fluoro-5-nitro-o-cresol
PC51193-1G
PC51193-250MG
PC51193-5G
APOSPC51193-1GEA
174
EUR
APOSPC51193-1G
APOSPC51193-250MG
APOSPC51193-5G
4-Fluoro-2-methyl-5-nitrophenol
4-Fluoro-2-methyl-5-nitrophenol
Formel:
C₇H₆FNO₃ Molecular Weight: 171,126 g/mol Schmelzpunkt: 138…142 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07779286 CAS-Nummer: 122455-84-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...