2-Bromo-6-methylphenyl trifluoromethanesulphonate 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Bromo-6-methylphenyl triflate, 3-Bromo-2-{[(trifluoromethyl)sulphonyl]oxy}toluene
PC51196-1G
PC51196-250MG
PC51196-5G
APOSPC51196-1GEA
140
EUR
APOSPC51196-1G
APOSPC51196-250MG
APOSPC51196-5G
2-Bromo-6-methylphenyl trifluoromethanesulphonate 95%
2-Bromo-6-methylphenyl trifluoromethanesulphonate
Formel:
C₈H₆BrF₃O₃S Molecular Weight: 319,096 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD27975433 CAS-Nummer: 1820716-91-3 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...