3-Bromo-6-chloro-2-fluoroaniline, N-BOC protected, tech

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromo-N-(tert-butoxycarbonyl)-6-chloro-2-fluoroaniline, tert-Butyl (3-bromo-6-chloro-2-fluorophenyl)carbamate

PC51198-1G PC51198-250MG
APOSPC51198-1GEA 847 EUR
APOSPC51198-1G APOSPC51198-250MG
3-Bromo-6-chloro-2-fluoroaniline, N-BOC protected, tech
3-Bromo-6-chloro-2-fluoroaniline, N-BOC protected
Formel: C₁₁H₁₂BrClFNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD28127019
CAS-Nummer: 1820711-67-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR