2-(Difluoromethyl)-4-hydroxypyridine 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Difluoromethyl)pyridin-4-ol, α,α-Difluoro-4-hydroxy-2-picoline

PC51200-1G PC51200-250MG
APOSPC51200-1GEA 723 EUR
APOSPC51200-1G APOSPC51200-250MG
2-(Difluoromethyl)-4-hydroxypyridine 95%
2-(Difluoromethyl)-4-hydroxypyridine
Formel: C₆H₅F₂NO
Molecular Weight: 145.107 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18820876
CAS-Nummer: 501125-75-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR