N-[(1R)-1-(4-Fluorophenyl)ethyl]cyclopropanamine hydrochloride

Lieferant: Apollo Scientific
Achtung

PC512022-1G PC512022-250MG
APOSPC512022-1GEA 208 EUR
APOSPC512022-1G APOSPC512022-250MG
N-[(1R)-1-(4-Fluorophenyl)ethyl]cyclopropanamine hydrochloride
N-[(1R)-1-(4-Fluorophenyl)ethyl]cyclopropanamine hydrochloride
Formel: C₁₁H₁₄FN·ClH
Molecular Weight: 215,695 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31714844
CAS-Nummer: 2366991-75-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR