N-[1-(2-Fluorophenyl)ethyl]cyclopropanamine hydrochloride

Lieferant: Apollo Scientific
Achtung

PC512024-1G PC512024-250MG PC512024-5G
APOSPC512024-1GEA 209 EUR
APOSPC512024-1G APOSPC512024-250MG APOSPC512024-5G
N-[1-(2-Fluorophenyl)ethyl]cyclopropanamine hydrochloride
N-[1-(2-Fluorophenyl)ethyl]cyclopropanamine hydrochloride
Formel: C₁₁H₁₄FN·ClH
Molecular Weight: 215,695 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12197062
CAS-Nummer: 1170249-65-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR