N-[1-(3-Fluorophenyl)ethyl]cyclopropanamine hydrochloride

Lieferant: Apollo Scientific
Achtung

PC512027-1G PC512027-250MG
APOSPC512027-1GEA 105 EUR
APOSPC512027-1G APOSPC512027-250MG
N-[1-(3-Fluorophenyl)ethyl]cyclopropanamine hydrochloride
N-[1-(3-Fluorophenyl)ethyl]cyclopropanamine hydrochloride
Formel: C₁₁H₁₄FN·ClH
Molecular Weight: 215,695 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD32066861
CAS-Nummer: 2379918-51-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR