4-Chloro-6-(trifluoromethyl)nicotinic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Chloro-6-(trifluoromethyl)pyridine-3-carboxylic acid, 5-Carboxy-4-chloro-2-(trifluoromethyl)pyridine
PC51210-1G
PC51210-250MG
PC51210-5G
APOSPC51210-1GEA
369
EUR
APOSPC51210-1G
APOSPC51210-250MG
APOSPC51210-5G
4-Chloro-6-(trifluoromethyl)nicotinic acid
4-Chloro-6-(trifluoromethyl)nicotinic acid
Formel:
C₇H₃ClF₃NO₂ Molecular Weight: 225,552 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13188812 CAS-Nummer: 1060810-66-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...