4-Hydroxy-5-iodo-2-(trifluoromethyl)pyridine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Hydroxy-5-iodo-α,α,α-trifluoro-2-picoline, 5-Iodo-2-(trifluoromethyl)pyridin-4-ol
PC51214-100MG
PC51214-1G
PC51214-250MG
APOSPC51214-100MGEA
140
EUR
APOSPC51214-100MG
APOSPC51214-1G
APOSPC51214-250MG
4-Hydroxy-5-iodo-2-(trifluoromethyl)pyridine
4-Hydroxy-5-iodo-2-(trifluoromethyl)pyridine
Formel:
C₆H₃F₃INO Lagertemperatur: Kühlschrank |
MDL:
MFCD24369747 CAS-Nummer: 1027818-89-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...