5-Amino-4-chloro-2-(trifluoromethyl)pyridine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Amino-4-chloro-α,α,α-trifluoro-2-picoline, 4-Chloro-6-(trifluoromethyl)pyridin-3-amine
PC51216-1G
PC51216-250MG
APOSPC51216-1GEA
549
EUR
APOSPC51216-1G
APOSPC51216-250MG
5-Amino-4-chloro-2-(trifluoromethyl)pyridine
5-Amino-4-chloro-2-(trifluoromethyl)pyridine
Formel:
C₆H₄ClF₃N₂ Molecular Weight: 196,558 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11848353 CAS-Nummer: 1196153-86-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...